CAS RN 400-70-4
IUPAC Name:
1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
Synonyms:
1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
259.99
Molecular Formula:
C7H2Cl2F3NO2
InChi Key:
VLVNHMVSVDVAOA-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=C(Cl)C=C1Cl)C(F)(F)F
1
–
1
of
1
results
1
2,4-Dichloro-5-nitrobenzotrifluoride, 97%, Thermo Scientific™
CAS: 400-70-4 Molecular Formula: C7H2Cl2F3NO2 Molecular Weight (g/mol): 259.993 MDL Number: MFCD00007073 InChI Key: VLVNHMVSVDVAOA-UHFFFAOYSA-N Synonym: 2,4-dichloro-5-nitrobenzotrifluoride,1,5-dichloro-2-nitro-4-trifluoromethyl benzene,2,4-dichloro-3,5-nitrobenzotrifluoride,benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl,2,4-dichloro-5-trifluoromethyl-1-nitrobenzene,pubchem4862,2,4-dichloro-5-nitro benzotrifluoride,3-trifluoromethyl-4,6-dichloronitrobenzene,2,4-dichloro-5-trifluoromethyl nitrobenzene PubChem CID: 621179 IUPAC Name: 1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene SMILES: C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F