accessibility menu, dialog, popup

UserName

CAS RN 39977-44-1

CH3OONOH

CAS RN 39977-44-1

IUPAC Name: methyl 6-(hydroxymethyl)pyridine-2-carboxylate
Synonyms: methyl 6-(hydroxymethyl)pyridine-2-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DVIUNMLAPDJWHL-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(CO)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results