accessibility menu, dialog, popup

UserName

CAS RN 399-36-0

CH3ONHFF

CAS RN 399-36-0

IUPAC Name: N-(2,4-difluorophenyl)acetamide
Synonyms: acetanilide,2,4-difluoro
Molecular Weight (g/mol): 171.15
Molecular Formula: C8H7F2NO
InChi Key: WOHLPEUHFSHZAN-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(F)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results