accessibility menu, dialog, popup

UserName

CAS RN 39489-79-7

IUPAC Name: 5-amino-2,4-dichlorophenol
Synonyms: 2-amino-4,6-dichlorophenol
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: QPHMVRPABQUYGN-UHFFFAOYSA-N
SMILES: NC1=CC(O)=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results