accessibility menu, dialog, popup

UserName

CAS RN 394-42-3

IUPAC Name: 2-fluoro-4-methoxybenzoate
Synonyms: 2-fluoro-4-methoxybenzoate
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H6FO3
InChi Key: UPWMPIKNUXTWFP-UHFFFAOYSA-M
SMILES: COC1=CC=C(C([O-])=O)C(F)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results