accessibility menu, dialog, popup

UserName

CAS RN 3938-95-2

H3COOCH3CH3H3C

CAS RN 3938-95-2

IUPAC Name: ethyl 2,2-dimethylpropanoate
Synonyms: ethyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: HHEIMYAXCOIQCJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results