accessibility menu, dialog, popup

UserName

OHOClFFF

CAS RN 39226-97-6

IUPAC Name: 2-chloro-3-(trifluoromethyl)benzoic acid
Synonyms: 2-chloro-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: AXOAWWUSRZCGKS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results