accessibility menu, dialog, popup

UserName

CAS RN 387358-52-3

NOBr

CAS RN 387358-52-3

IUPAC Name: 5-(2-bromophenyl)-1,2-oxazole
Synonyms: 5-(2-bromophenyl)-1,2-oxazole
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: CFIWXQDRNFECMZ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=NO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results