accessibility menu, dialog, popup

UserName

KH3COOOO

CAS RN 38330-80-2

IUPAC Name: potassium 3-methoxy-3-oxopropanoate
Synonyms: potassium 3-methoxy-3-oxopropanoate
Molecular Weight (g/mol): 156.18
Molecular Formula: C4H5KO4
InChi Key: WWTULTKUWBKVGV-UHFFFAOYSA-M
SMILES: [K+].COC(=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results