CAS RN 38284-47-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S,2S,5S)-4-[2-[2-(6,6-Dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]morpholine, TRC
CAS: 38284-47-8 Molecular Formula: C17H31NO2 Molecular Weight (g/mol): 281.43 Synonym: (1S,2S,5S)-2-[2-(2-Morpholinoethoxy)ethyl]-6,6-dimethylnorpinane,(1S,2S,5S)-6,6-Dimethyl-2-[2-(2-morpholinoethoxy)ethyl]norpinane; IUPAC Name: 4-[2-[2-(6,6-dimethyl-4-bicyclo[3.1.1]heptanyl)ethoxy]ethyl]morpholine SMILES: CC1(C)[C@@H]2[C@@H](CCOCCN3CCOCC3)CC[C@H]1C2