accessibility menu, dialog, popup

UserName

CAS RN 38186-82-2

CH3NClNH2

CAS RN 38186-82-2

IUPAC Name: 6-chloro-5-methylpyridin-3-amine
Synonyms: 6-chloro-5-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: VSBISZPNLZFTPG-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results