CAS RN 375368-80-2
CAS RN 375368-80-2
IUPAC Name:
3-fluoro-2-methoxy-6-methylpyridine
Synonyms:
3-fluoro-2-methoxy-6-methylpyridine
Molecular Weight (g/mol):
141.15
Molecular Formula:
C7H8FNO
InChi Key:
DAQMWRIPXIPMAA-UHFFFAOYSA-N
SMILES:
COC1=C(F)C=CC(C)=N1
1
–
1
of
1
results
1
3-Fluoro-2-methoxy-6-methylpyridine, 97%, Thermo Scientific Chemicals
CAS: 375368-80-2 Molecular Formula: C7H8FNO Molecular Weight (g/mol): 141.15 MDL Number: MFCD03095083 InChI Key: DAQMWRIPXIPMAA-UHFFFAOYSA-N Synonym: 3-fluoro-2-methoxy-6-picoline,2-methoxy-3-fluoro-6-methylpyridine,3-fluoro-2-methoxy-6-methyl-pyridine,pubchem6585,ksc914i9l,abbypharma ap-15-5217,pyridine, 3-fluoro-2-methoxy-6-methyl,pyridine, 3-fluoro-2-methoxy-6-methyl-9ci PubChem CID: 24820319 IUPAC Name: 3-fluoro-2-methoxy-6-methylpyridine SMILES: COC1=C(F)C=CC(C)=N1