CAS RN 372183-68-1
CAS RN 372183-68-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(3-Fluorophenoxy)ethylamine hydrochloride, 98%, Thermo Scientific™
CAS: 372183-68-1 Molecular Formula: C8H10FNO Molecular Weight (g/mol): 155.172 MDL Number: MFCD11099553 InChI Key: UTSSEDORJLCBMS-UHFFFAOYSA-N Synonym: 2-3-fluorophenoxy ethylamine,2-3-fluorophenoxy ethanamine,2-3-fluoro-phenoxy-ethylamine,1-2-aminoethoxy-3-fluorobenzene,ethanamine,2-3-fluorophenoxy,2-3-fluorophenoxy ethan-1-amine,ethanamine, 2-3-fluorophenoxy,pubchem19158,acmc-1cfl9 PubChem CID: 14245330 IUPAC Name: 2-(3-fluorophenoxy)ethanamine SMILES: C1=CC(=CC(=C1)F)OCCN