accessibility menu, dialog, popup

UserName

CH3OONClCH3

CAS RN 37041-32-0

IUPAC Name: ethyl 4-chloro-8-methylquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-8-methylquinoline-3-carboxylate
Molecular Weight (g/mol): 249.69
Molecular Formula: C13H12ClNO2
InChi Key: PGGVUZUNDXCMSS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(C)=CC=CC2=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results