CAS RN 367-30-6
IUPAC Name:
2,5-difluoroaniline
Synonyms:
2,5-difluoroaniline
Molecular Weight (g/mol):
129.11
Molecular Formula:
C6H5F2N
InChi Key:
YNOOQIUSYGWMSS-UHFFFAOYSA-N
SMILES:
NC1=CC(F)=CC=C1F
1
–
1
of
1
results
1
2,5-Difluoroaniline, 98%
CAS: 367-30-6 Molecular Formula: C6H5F2N Molecular Weight (g/mol): 129.11 MDL Number: MFCD00007651 InChI Key: YNOOQIUSYGWMSS-UHFFFAOYSA-N Synonym: benzenamine, 2,5-difluoro,2,5-difluorobenzenamine,2,5-difluoro-phenylamine,aniline, 2,5-difluoro,2.5-difluoroaniline,aniline,5-difluoro,pubchem1531,2,5-difluoro-aniline,2,5-difluorophenylamine,benzenamine,5-difluoro PubChem CID: 67775 IUPAC Name: 2,5-difluoroaniline SMILES: C1=CC(=C(C=C1F)N)F