accessibility menu, dialog, popup

UserName

CAS RN 36476-86-5

NN

CAS RN 36476-86-5

IUPAC Name: 1-(diphenylmethyl)azetidine-3-carbonitrile
Synonyms: 1-(diphenylmethyl)azetidine-3-carbonitrile
Molecular Weight (g/mol): 248.33
Molecular Formula: C17H16N2
InChi Key: IXMOEAHDRKNAAG-UHFFFAOYSA-N
SMILES: N#CC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results