accessibility menu, dialog, popup

UserName

CAS RN 36282-40-3

OH3CMgBr

CAS RN 36282-40-3

IUPAC Name: magnesium(2+) anisole bromide
Synonyms: magnesium(2+) anisole bromide
Molecular Weight (g/mol): 211.34
Molecular Formula: C7H7BrMgO
InChi Key: FKUUDDGRDRPAQQ-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].COC1=C[C-]=CC=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)