accessibility menu, dialog, popup

UserName

CAS RN 3609-87-8

BrSOOHN

CAS RN 3609-87-8

IUPAC Name: N-benzyl-4-bromobenzene-1-sulfonamide
Synonyms: N-benzyl-4-bromobenzenesulfonamide
Molecular Weight (g/mol): 326.21
Molecular Formula: C13H12BrNO2S
InChi Key: KBFYMIDQZWIKJR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)S(=O)(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results