accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFF

CAS RN 355-37-3

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
Molecular Weight (g/mol): 320.05
Molecular Formula: C6HF13
InChi Key: XJSRKJAHJGCPGC-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)