accessibility menu, dialog, popup

UserName

CAS RN 35288-49-4

IUPAC Name: 3-[4-(3-chloro-3-oxoprop-1-en-1-yl)phenyl]prop-2-enoyl chloride
Synonyms: 3-[4-(3-chloro-3-oxoprop-1-en-1-yl)phenyl]prop-2-enoyl chloride
Molecular Weight (g/mol): 255.09
Molecular Formula: C12H8Cl2O2
InChi Key: WQZLRZFBCIUDFL-UHFFFAOYSA-N
SMILES: ClC(=O)C=CC1=CC=C(C=CC(Cl)=O)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results