accessibility menu, dialog, popup

UserName

CAS RN 3512-17-2

FFNF

CAS RN 3512-17-2

IUPAC Name: 2,4,6-trifluoropyridine
Synonyms: 2,4,6-trifluoropyridine
Molecular Weight (g/mol): 133.07
Molecular Formula: C5H2F3N
InChi Key: UZDRWXKBKVVUTE-UHFFFAOYSA-N
SMILES: FC1=CC(F)=NC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results