CAS RN 3496-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyanomelamine, TRC
CAS: 3496-98-8 Molecular Formula: C4 H5 N7 Molecular Weight (g/mol): 151.13 Synonym: 2-(Cyanoamino)-4,6-diamino-sym-triazine,4,6-Diamino-s-triazine-2-carbamonitrile,(4,6-Diamino-1,3,5-triazin-2-yl)cyanamide,N-(4,6-diamino-1,3,5-triazin-2-yl)cyanamide IUPAC Name: (4,6-diamino-1,3,5-triazin-2-yl)cyanamide SMILES: Nc1nc(N)nc(NC#N)n1