accessibility menu, dialog, popup

UserName

CAS RN 345909-31-1

OOOOCaH3CH3CCH3CH3

CAS RN 345909-31-1

IUPAC Name: calcium bis(4-oxopent-2-en-2-olate) hydrate
Synonyms: calcium bis(4-oxopent-2-en-2-olate) hydrate
Molecular Weight (g/mol): 256.31
Molecular Formula: C10H16CaO5
InChi Key: FFWWBMKQRYMOKA-UHFFFAOYSA-L
SMILES: O.[Ca++].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results