accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 3453-33-6

IUPAC Name: 6-methoxynaphthalene-2-carbaldehyde
Synonyms: 6-methoxynaphthalene-2-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: VZBLASFLFFMMCM-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C=O)C=CC2=C1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results