accessibility menu, dialog, popup

UserName

CAS RN 34452-51-2

KOOHS

CAS RN 34452-51-2

IUPAC Name: potassium 2-sulfanylacetate
Synonyms: potassium thioglycolate
Molecular Weight (g/mol): 130.2
Molecular Formula: C2H3KO2S
InChi Key: HYTYHTSMCRDHIM-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CS
Percent Purity 
  • (1)
Quantity 
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)