accessibility menu, dialog, popup

UserName

CAS RN 34404-32-5

NH2(R)HNOOOHO

CAS RN 34404-32-5

IUPAC Name: 2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: 2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: CKGCFBNYQJDIGS-UHFFFAOYNA-N
SMILES: NC(CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)