CAS RN 342406-26-2
CAS RN 342406-26-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bismuth subgallate hydrate
CAS: 342406-26-2 Molecular Formula: C7H5BiO6 Molecular Weight (g/mol): 394.09 MDL Number: MFCD00044980 InChI Key: JAONZGLTYYUPCT-UHFFFAOYSA-K Synonym: bismuth subgallate,wismutgallathydroxid,dermatol,bismuth gallate,dermatol puder,bismuth subgallas,bismutum subgallicum,basic bismuth gallate,gallic acid bismuth basic salt,wismutgallat, basisches PubChem CID: 16682999 IUPAC Name: 4-hydroxy-1,3,2$l^{2}-benzodioxabismole-6-carboxylic acid;hydrate SMILES: O[Bi]1OC2=C(O1)C(O)=CC(=C2)C(O)=O