accessibility menu, dialog, popup

UserName

CAS RN 3377-86-4

IUPAC Name: 2-bromohexane
Synonyms: 2-bromohexane
Molecular Weight (g/mol): 165.07
Molecular Formula: C6H13Br
InChi Key: NEBYCXAKZCQWAW-UHFFFAOYNA-N
SMILES: CCCCC(C)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results