accessibility menu, dialog, popup

UserName

CAS RN 33588-54-4

IUPAC Name: 1-acetyl-5-bromo-1H-indol-3-yl acetate
Synonyms: 1-acetyl-5-bromoindol-3-yl acetate
Molecular Weight (g/mol): 296.12
Molecular Formula: C12H10BrNO3
InChi Key: XJRIDJAGAYGJCK-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CN(C(C)=O)C2=CC=C(Br)C=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results