accessibility menu, dialog, popup

UserName

CAS RN 3289-28-9

H3COO

CAS RN 3289-28-9

IUPAC Name: ethyl cyclohexanecarboxylate
Synonyms: cyclohexanecarboxylic acid, ethyl ester
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: JJOYCHKVKWDMEA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results