accessibility menu, dialog, popup

UserName

CAS RN 324-27-6

IUPAC Name: 2-(4-fluorophenyl)-1H-1,3-benzodiazole
Synonyms: 2-(4-fluorophenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 212.23
Molecular Formula: C13H9FN2
InChi Key: FPWUSPPQEHBWHC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results