accessibility menu, dialog, popup

UserName

H3CNCH3OH

CAS RN 3179-63-3

IUPAC Name: 3-(dimethylamino)propan-1-ol
Synonyms: 1-propanol, 3-(dimethylamino)-
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: PYSGFFTXMUWEOT-UHFFFAOYSA-N
SMILES: CN(C)CCCO
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)