accessibility menu, dialog, popup

UserName

HClCH3ClH2N

CAS RN 3165-93-3

IUPAC Name: hydrogen 4-chloro-2-methylaniline chloride
Synonyms: hydrogen 4-chloro-o-toluidine chloride
Molecular Weight (g/mol): 178.06
Molecular Formula: C7H9Cl2N
InChi Key: VKYZDCTWJGBFDW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(Cl)=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results