accessibility menu, dialog, popup

UserName

CAS RN 311346-69-7

H3COONFFCl

CAS RN 311346-69-7

IUPAC Name: ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
Molecular Weight (g/mol): 271.65
Molecular Formula: C12H8ClF2NO2
InChi Key: IEGYWXRVOVIAMV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=C(F)C=C(F)C2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results