accessibility menu, dialog, popup

UserName

BFFFFCH3NNH3CH3C

CAS RN 307492-75-7

IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 212
Molecular Formula: C7H13BF4N2
InChi Key: KNIWGPCMWBGRBE-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCN1C=C[N+](C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results