CAS RN 3056-44-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Gluco-Hexodialdose, TRC
CAS: 3056-44-8 Molecular Formula: C6H10O6 Molecular Weight (g/mol): 178.14 Synonym: Glucodialdose,L-Gulo-Hexodialdose; IUPAC Name: (2S,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanedial SMILES: O[C@H](C=O)[C@@H](O)[C@H](O)[C@@H](O)C=O