accessibility menu, dialog, popup

UserName

CAS RN 304655-78-5

NNHNNH2OOH

CAS RN 304655-78-5

IUPAC Name: 3-amino-1H-1,2,4-triazole-5-carboxylic acid hydrate
Synonyms: 5-amino-2H-1,2,4-triazole-3-carboxylic acid hydrate
Molecular Weight (g/mol): 146.11
Molecular Formula: C3H6N4O3
InChi Key: AGJCUQSNRFXRDU-UHFFFAOYSA-N
SMILES: O.NC1=NNC(=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results