CAS RN 29621-88-3
CAS RN 29621-88-3
IUPAC Name:
(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]diselanyl}propanoic acid
Synonyms:
(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]diselanyl}propanoic acid
Molecular Weight (g/mol):
334.11
Molecular Formula:
C6H12N2O4Se2
InChi Key:
JULROCUWKLNBSN-IMJSIDKUSA-N
SMILES:
N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O
1
–
1
of
1
results
1
L-Selenocystine, 95%, Thermo Scientific Chemicals
CAS: 29621-88-3 Molecular Formula: C6H12N2O4Se2 Molecular Weight (g/mol): 334.11 MDL Number: MFCD00800971 InChI Key: JULROCUWKLNBSN-IMJSIDKUSA-N Synonym: l-selenocystine,seleno-l-cystine,l-3,3'-diselenodialanine,alanine, 3,3'-diselenodi-, l,selenocystine, l,l-alanine,3,3'-diselenobis,2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid,l-selenocystine monohydrate,selenocystine, l-isomer PubChem CID: 207306 IUPAC Name: (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid SMILES: N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O