CAS RN 295-14-7
CAS RN 295-14-7
IUPAC Name:
1,4,7,10-tetraazacyclotridecane
Synonyms:
1,4,7,10-tetraazacyclotridecane
Molecular Weight (g/mol):
186.3
Molecular Formula:
C9H22N4
InChi Key:
LADZJJOUGVGJHM-UHFFFAOYSA-N
SMILES:
C1CNCCNCCNCCNC1
1
–
1
of
1
results
1
1,4,7,10-Tetraazacyclotridecane
CAS: 295-14-7 Molecular Formula: C9H22N4 Molecular Weight (g/mol): 186.303 MDL Number: MFCD06410986 InChI Key: LADZJJOUGVGJHM-UHFFFAOYSA-N Synonym: 1,4,7,10-tetraazacyclotridecane,homocyclen,acmc-20ap3n,13 anen4 PubChem CID: 11478723 IUPAC Name: 1,4,7,10-tetrazacyclotridecane SMILES: C1CNCCNCCNCCNC1