accessibility menu, dialog, popup

UserName

CAS RN 29270-30-2

OOHBrCl

CAS RN 29270-30-2

IUPAC Name: 2-bromo-2-(2-chlorophenyl)acetic acid
Synonyms: bromo(2-chlorophenyl)acetic acid
Molecular Weight (g/mol): 249.49
Molecular Formula: C8H6BrClO2
InChi Key: XHAPROULWZYBGA-UHFFFAOYNA-N
SMILES: OC(=O)C(Br)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)