CAS RN 290296-72-9
CAS RN 290296-72-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Netupitant metabolite N-desmethyl Netupitant, MedChemExpress
MedChemExpress N-desmethyl Netupitant is a metabolite of Netupitant, which is an antiemitic drug.