accessibility menu, dialog, popup

UserName

H3COOClClOOCH3

CAS RN 28751-26-0

IUPAC Name: 1,3-diethyl 2-(3,4-dichlorophenyl)propanedioate
Synonyms: 1,3-diethyl 2-(3,4-dichlorophenyl)propanedioate
Molecular Weight (g/mol): 305.15
Molecular Formula: C13H14Cl2O4
InChi Key: GZSMDWSQPZMQDV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C1=CC(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results