CAS RN 28598-08-5
CAS RN 28598-08-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cinoctramide, MedChemExpress
MedChemExpress Cinoctramide is an intermediate of pharmaceutical synthesis. Cinoctramide can be used for the research of autoimmune diseases.