accessibility menu, dialog, popup

UserName

CAS RN 278610-39-2

NClNOO

CAS RN 278610-39-2

IUPAC Name: 1-(2-chloropyridin-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(2-chloropyridin-3-yl)pyrrole-2,5-dione
Molecular Weight (g/mol): 208.6
Molecular Formula: C9H5ClN2O2
InChi Key: JVJJPDZQLVCNFU-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC=N1)N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results