accessibility menu, dialog, popup

UserName

CAS RN 2689-69-2

IUPAC Name: methyl 3-oxothiolane-2-carboxylate
Synonyms: methyl 3-oxothiolane-2-carboxylate
Molecular Weight (g/mol): 160.19
Molecular Formula: C6H8O3S
InChi Key: TUSSXVYKOOVOID-UHFFFAOYNA-N
SMILES: COC(=O)C1SCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results