accessibility menu, dialog, popup

UserName

CAS RN 26820-62-2

ONONNO

CAS RN 26820-62-2

IUPAC Name: 4-(5-nitropyridin-2-yl)morpholine
Synonyms: 4-(5-nitropyridin-2-yl)morpholine
Molecular Weight (g/mol): 209.21
Molecular Formula: C9H11N3O3
InChi Key: FYJXIPHXFGFEGT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results