CAS RN 261763-25-1
IUPAC Name:
2-[5-chloro-2-(trifluoromethyl)phenyl]acetic acid
Synonyms:
[5-chloro-2-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
238.59
Molecular Formula:
C9H6ClF3O2
InChi Key:
GINCGCMLTQDEJC-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=C(C=CC(Cl)=C1)C(F)(F)F
1
–
1
of
1
results
1
5-Chloro-2-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
CAS: 261763-25-1 Molecular Formula: C9H6ClF3O2 Molecular Weight (g/mol): 238.59 MDL Number: MFCD01631358 InChI Key: GINCGCMLTQDEJC-UHFFFAOYSA-N Synonym: 5-chloro-2-trifluoromethyl phenylacetic acid,2-5-chloro-2-trifluoromethyl phenyl acetic acid,5-chloro-2-trifluoromethyl-phenyl-acetic acid,5-chloro-2-trifluoromethyl phenyl acetic acid,pubchem4311,ksc495g3b,5-chloro-2-trifluoromethylphenylacetic acid PubChem CID: 2778122 IUPAC Name: 2-[5-chloro-2-(trifluoromethyl)phenyl]acetic acid SMILES: C1=CC(=C(C=C1Cl)CC(=O)O)C(F)(F)F