accessibility menu, dialog, popup

UserName

CAS RN 2613-22-1

OHClF

CAS RN 2613-22-1

IUPAC Name: 3-chloro-2-fluorophenol
Synonyms: 3-chloro-2-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: PCHPYNHSMSAJEU-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results