accessibility menu, dialog, popup

UserName

CAS RN 25658-42-8

NZr

CAS RN 25658-42-8

IUPAC Name: zirconioamine
Synonyms: zirconium nitride
Molecular Weight (g/mol): 105.23
Molecular Formula: NZr
InChi Key: ZVWKZXLXHLZXLS-UHFFFAOYSA-N
SMILES: N#[Zr]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results