accessibility menu, dialog, popup

UserName

CAS RN 256417-11-5

H3COOFF

CAS RN 256417-11-5

IUPAC Name: 2,3-difluoro-4-methoxybenzaldehyde
Synonyms: 2,3-difluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: UBBBLLSKHCENQZ-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results